accessibility menu, dialog, popup

UserName

CAS RN 133407-83-7

IUPAC Name: benzyl N-[3-methyl-1-({3-methyl-1-[(1-oxohexan-2-yl)carbamoyl]butyl}carbamoyl)butyl]carbamate
Synonyms: benzyl N-[3-methyl-1-({3-methyl-1-[(1-oxohexan-2-yl)carbamoyl]butyl}carbamoyl)butyl]carbamate
Molecular Weight (g/mol): 475.63
Molecular Formula: C26H41N3O5
InChi Key: RNPDUXVFGTULLP-UHFFFAOYNA-N
SMILES: CCCCC(NC(=O)C(CC(C)C)NC(=O)C(CC(C)C)NC(=O)OCC1=CC=CC=C1)C=O
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results