accessibility menu, dialog, popup

UserName

OHOFFF

CAS RN 133261-33-3

IUPAC Name: 4-(trifluoromethyl)cyclohexane-1-carboxylic acid
Synonyms: 4-(trifluoromethyl)cyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 196.17
Molecular Formula: C8H11F3O2
InChi Key: LMEAZIIFLVDISW-UHFFFAOYSA-N
SMILES: OC(=O)C1CCC(CC1)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)