CAS RN 133053-19-7
CAS RN 133053-19-7
IUPAC Name:
3-{1-[3-(dimethylamino)propyl]-5-methoxy-1H-indol-3-yl}-4-(1H-indol-3-yl)-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms:
3-{1-[3-(dimethylamino)propyl]-5-methoxyindol-3-yl}-4-(1H-indol-3-yl)-1H-pyrrole-2,5-dione
Molecular Weight (g/mol):
442.52
Molecular Formula:
C26H26N4O3
InChi Key:
LLJJDLHGZUOMQP-UHFFFAOYSA-N
SMILES:
COC1=CC=C2N(CCCN(C)C)C=C(C2=C1)C1=C(C(=O)NC1=O)C1=CNC2=CC=CC=C12
1
–
1
of
1
results
1
Thermo Scientific Chemicals Go 6983, 98%
CAS: 133053-19-7 Molecular Formula: C26H26N4O3 Synonym: 3-[1-[3-(dimethylamino)propyl]-5-methoxy-1H-indol-3-yl]-4-(1H-indol-3-yl)-1H-pyrrole-2,5-dione;