accessibility menu, dialog, popup

UserName

H3CNOONH2HCl

CAS RN 132873-57-5

IUPAC Name: 1-(4-nitrophenyl)ethan-1-amine hydrochloride
Synonyms: 1-(4-nitrophenyl)ethanamine hydrochloride
Molecular Weight (g/mol): 202.64
Molecular Formula: C8H11ClN2O2
InChi Key: CZQQGVFHLSBEDV-UHFFFAOYNA-N
SMILES: Cl.CC(N)C1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results