accessibility menu, dialog, popup

UserName

CAS RN 13265-84-4

IUPAC Name: (2R,4R)-2-(hydroxymethyl)-3,4-dihydro-2H-pyran-3,4-diol
Synonyms: glucal
Molecular Weight (g/mol): 146.14
Molecular Formula: C6H10O4
InChi Key: YVECGMZCTULTIS-QYRBDRAASA-N
SMILES: OC[C@H]1OC=C[C@@H](O)C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results