accessibility menu, dialog, popup

UserName

CH3NClBr

CAS RN 132606-40-7

IUPAC Name: 3-bromo-6-chloro-2-methylpyridine
Synonyms: 3-bromo-6-chloro-2-methylpyridine
Molecular Weight (g/mol): 206.47
Molecular Formula: C6H5BrClN
InChi Key: DTFBHJWQTDQBEM-UHFFFAOYSA-N
SMILES: CC1=C(Br)C=CC(Cl)=N1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)