accessibility menu, dialog, popup

UserName

CH3OONNO

CAS RN 132546-81-7

IUPAC Name: methyl 6-(morpholin-4-yl)pyridine-3-carboxylate
Synonyms: methyl 6-(morpholin-4-yl)pyridine-3-carboxylate
Molecular Weight (g/mol): 222.24
Molecular Formula: C11H14N2O3
InChi Key: FFBPRHFKXVNLDV-UHFFFAOYSA-N
SMILES: COC(=O)C1=CN=C(C=C1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)