accessibility menu, dialog, popup

UserName

ClONHNO

CAS RN 13214-53-4

IUPAC Name: 2-oxoimidazolidine-1-carbonyl chloride
Synonyms: 2-oxoimidazolidine-1-carbonyl chloride
Molecular Weight (g/mol): 148.55
Molecular Formula: C4H5ClN2O2
InChi Key: NXJZQSRAFBHNLI-UHFFFAOYSA-N
SMILES: ClC(=O)N1CCNC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results