accessibility menu, dialog, popup

UserName

CH3H3CCH3OOHN(S)NOOOHO

CAS RN 131980-29-5

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(3-nitrophenyl)propanoic acid
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-3-(3-nitrophenyl)propanoic acid
Molecular Weight (g/mol): 310.31
Molecular Formula: C14H18N2O6
InChi Key: OWTGPXDXLMNQKK-NSHDSACASA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CC1=CC=CC(=C1)[N+]([O-])=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results