CAS RN 1318-93-0
IUPAC Name:
dialuminium(3+) tetrakis(silanedione) hydrate trioxidandiide
Synonyms:
montmorillonite
Molecular Weight (g/mol):
360.31
Molecular Formula:
Al2H2O12Si4
InChi Key:
GUJOJGAPFQRJSV-UHFFFAOYSA-N
SMILES:
O.[O--].[O--].[O--].[Al+3].[Al+3].O=[Si]=O.O=[Si]=O.O=[Si]=O.O=[Si]=O
1
–
1
of
1
results
1
Montmorillonite K10
CAS: 1318-93-0 Molecular Formula: Al2H2O12Si4 Molecular Weight (g/mol): 360.307 MDL Number: MFCD00132796 InChI Key: GUJOJGAPFQRJSV-UHFFFAOYSA-N Synonym: montmorillonite,bentolite,gelwhite l,montmorillonite ksf,montmorillonite k 10,unii-a585mn1h2l,envirobent,neokunibond,arcillite,bedelix PubChem CID: 71586775 IUPAC Name: dialuminum;dioxosilane;oxygen(2-);hydrate SMILES: O.[O-2].[O-2].[O-2].O=[Si]=O.O=[Si]=O.O=[Si]=O.O=[Si]=O.[Al+3].[Al+3]