accessibility menu, dialog, popup

UserName

OTaOTaOOO

CAS RN 1314-61-0

IUPAC Name: [(dioxotantalio)oxy]dioxotantalum
Synonyms: tantalum pentoxide
Molecular Weight (g/mol): 441.89
Molecular Formula: O5Ta2
InChi Key: PBCFLUZVCVVTBY-UHFFFAOYSA-N
SMILES: O=[Ta](=O)O[Ta](=O)=O
Grade 
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (7)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)