CAS RN 13138-48-2
CAS RN 13138-48-2
IUPAC Name:
3,3'-dimethyl-[1,1'-binaphthalene]-4,4'-diamine
Synonyms:
3,3'-dimethyl-[1,1'-binaphthalene]-4,4'-diamine
Molecular Weight (g/mol):
312.42
Molecular Formula:
C22H20N2
InChi Key:
FTEGMIZQLGXBNA-UHFFFAOYSA-N
SMILES:
CC1=CC(=C2C=CC=CC2=C1N)C1=C2C=CC=CC2=C(N)C(C)=C1
1
–
1
of
1
results
1
3,3'-Dimethylnaphthidine 97.0+%, TCI America™
CAS: 13138-48-2 Molecular Formula: C22H20N2 Molecular Weight (g/mol): 312.42 MDL Number: MFCD00042706 InChI Key: FTEGMIZQLGXBNA-UHFFFAOYSA-N PubChem CID: 83166 IUPAC Name: 3,3'-dimethyl-[1,1'-binaphthalene]-4,4'-diamine SMILES: CC1=CC(=C2C=CC=CC2=C1N)C1=C2C=CC=CC2=C(N)C(C)=C1