accessibility menu, dialog, popup

UserName

KSiOOOK

CAS RN 1312-76-1

IUPAC Name: dipotassium oxosilanebis(olate)
Synonyms: potassium silicate
Molecular Weight (g/mol): 154.28
Molecular Formula: K2O3Si
InChi Key: NNHHDJVEYQHLHG-UHFFFAOYSA-N
SMILES: [K+].[K+].[O-][Si]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results