CAS RN 131182-68-8
IUPAC Name:
methoxy[4-(methoxydimethylsilyl)phenyl]dimethylsilane
Synonyms:
methoxy[4-(methoxydimethylsilyl)phenyl]dimethylsilane
Molecular Weight (g/mol):
254.48
Molecular Formula:
C12H22O2Si2
InChi Key:
VQECZZRUEDMVCU-UHFFFAOYSA-N
SMILES:
CO[Si](C)(C)C1=CC=C(C=C1)[Si](C)(C)OC
1
–
1
of
1
results
1
Promotions Available
1,4-Bis(methoxydimethylsilyl)benzene, 97%
1,4-Bis(methoxydimethylsilyl)benzene, 97%
CAS: 131182-68-8 Molecular Formula: C12H22O2Si2 Molecular Weight (g/mol): 254.476 MDL Number: MFCD09266162 InChI Key: VQECZZRUEDMVCU-UHFFFAOYSA-N Synonym: 1,4-bis methoxydimethylsilyl benzene,methoxy 4-methoxydimethylsilyl phenyl dimethylsilane,amtsi067,1,4-phenylenebis methoxydimethylsilane,p-bis dimethylmethoxysilyl benzene,1,4-phenylene bis methoxy dimethyl silane,methoxy-4-methoxy dimethyl silyl phenyl-dimethylsilane PubChem CID: 21900341 IUPAC Name: methoxy-[4-[methoxy(dimethyl)silyl]phenyl]-dimethylsilane SMILES: CO[Si](C)(C)C1=CC=C(C=C1)[Si](C)(C)OC