accessibility menu, dialog, popup

UserName

H2NONH2O

CAS RN 131-14-6

IUPAC Name: 2,6-diamino-9,10-dihydroanthracene-9,10-dione
Synonyms: 2,6-diaminoanthracene-9,10-dione
Molecular Weight (g/mol): 238.25
Molecular Formula: C14H10N2O2
InChi Key: WQOWBWVMZPPPGX-UHFFFAOYSA-N
SMILES: NC1=CC=C2C(=O)C3=CC(N)=CC=C3C(=O)C2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results