CAS RN 13083-04-0
IUPAC Name:
cerium(3+) bis(nitric acid) diamine tetrahydrate trinitrate
Synonyms:
cerium(3+) bis(nitric acid) diamine tetrahydrate trinitrate
Molecular Weight (g/mol):
558.27
Molecular Formula:
CeH16N7O19
InChi Key:
AIVZRMAXALRCHB-UHFFFAOYSA-N
SMILES:
N.N.O.O.O.O.[Ce+3].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
1
–
1
of
1
results
1
Cerium(III) ammonium nitrate tetrahydrate, Reagent Grade
CAS: 13083-04-0 Molecular Formula: CeH16N7O19 Molecular Weight (g/mol): 558.27 MDL Number: MFCD00213483 InChI Key: AIVZRMAXALRCHB-UHFFFAOYSA-N Synonym: ce.3no3.2hno3.4h2o.2nh3,ammonium cerium iii nitrate tetrahydrate,cerous ammonium nitrate, hydrated, reagent,cerium iii ammonium nitrate tetrahydrate, reagent grade,cerium 3+ bis nitric acid diamine tetrahydrate trinitrate PubChem CID: 91886599 IUPAC Name: azane;cerium(3+);nitric acid;trinitrate;tetrahydrate SMILES: N.N.O.O.O.O.[Ce+3].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O