accessibility menu, dialog, popup

UserName

H3CNCH3H3CH3CClBrCl

CAS RN 13053-75-3

IUPAC Name: dichlorobromanuide; tetrabutylazanium
Synonyms: dichlorobromanuide; tetrabutylammonium ion
Molecular Weight (g/mol): 393.28
Molecular Formula: C16H36BrCl2N
InChi Key: XKVIJLXGNLMYCT-UHFFFAOYSA-N
SMILES: Cl[Br-]Cl.CCCC[N+](CCCC)(CCCC)CCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results