accessibility menu, dialog, popup

UserName

CAS RN 1295-35-8

Ni

CAS RN 1295-35-8

IUPAC Name: bis((1Z,5Z)-cycloocta-1,5-diene) nickel
Synonyms: bis(cyclooctadiene)nickel(0)
Molecular Weight (g/mol): 275.06
Molecular Formula: C16H24Ni
InChi Key: JRTIUDXYIUKIIE-KZUMESAESA-N
SMILES: [Ni].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)