accessibility menu, dialog, popup

UserName

NNHNN

CAS RN 129485-83-2

IUPAC Name: 2-[3-(pyridin-2-yl)-1H-pyrazol-5-yl]pyridine
Synonyms: 2-[5-(pyridin-2-yl)-2H-pyrazol-3-yl]pyridine
Molecular Weight (g/mol): 222.25
Molecular Formula: C13H10N4
InChi Key: IMDRKCUYKQQEAC-UHFFFAOYSA-N
SMILES: N1N=C(C=C1C1=CC=CC=N1)C1=CC=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results