accessibility menu, dialog, popup

UserName

CAS RN 129136-65-8

IUPAC Name: 4-[(thiophen-2-yl)methyl]aniline
Synonyms: 4-(thiophen-2-ylmethyl)aniline
Molecular Weight (g/mol): 189.28
Molecular Formula: C11H11NS
InChi Key: PIJZLXNJWCKMSA-UHFFFAOYSA-N
SMILES: NC1=CC=C(CC2=CC=CS2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)