accessibility menu, dialog, popup

UserName

H3CNHNHH3COO

CAS RN 128-80-3

IUPAC Name: 1,4-bis[(4-methylphenyl)amino]-9,10-dihydroanthracene-9,10-dione
Synonyms: quinizarin green SS quinizarine green SS
Molecular Weight (g/mol): 418.5
Molecular Formula: C28H22N2O2
InChi Key: TVRGPOFMYCMNRB-UHFFFAOYSA-N
SMILES: CC1=CC=C(NC2=CC=C(NC3=CC=C(C)C=C3)C3=C2C(=O)C2=CC=CC=C2C3=O)C=C1
Color 
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results