accessibility menu, dialog, popup

UserName

CAS RN 1273-73-0

IUPAC Name: Bromoferrocene
Synonyms: —
Molecular Weight (g/mol): 264.93
Molecular Formula: C10H9BrFe
InChi Key: —
SMILES: [Fe].c1cccc1.Brc1cccc1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results