accessibility menu, dialog, popup

UserName

NaONOSOOO

CAS RN 127-68-4

IUPAC Name: sodium 3-nitrobenzene-1-sulfonate
Synonyms: sodium M-nitrobenzenesulfonate
Molecular Weight (g/mol): 225.15
Molecular Formula: C6H4NNaO5S
InChi Key: LJRGBERXYNQPJI-UHFFFAOYSA-M
SMILES: [Na+].[O-][N+](=O)C1=CC=CC(=C1)S([O-])(=O)=O
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)