CAS RN 126533-97-9
IUPAC Name:
2-(morpholin-4-yl)-1,3-thiazole-4-carbaldehyde
Synonyms:
2-(morpholin-4-yl)-1,3-thiazole-4-carbaldehyde
Molecular Weight (g/mol):
198.24
Molecular Formula:
C8H10N2O2S
InChi Key:
JPJSUXPYPAQDDL-UHFFFAOYSA-N
SMILES:
O=CC1=CSC(=N1)N1CCOCC1
1
–
2
of
2
results
2
2-Morpholino-1,3-thiazole-4-carbaldehyde, 97%, Thermo Scientific™
CAS: 126533-97-9 Molecular Formula: C8H10N2O2S Molecular Weight (g/mol): 198.24 MDL Number: MFCD09817537 InChI Key: JPJSUXPYPAQDDL-UHFFFAOYSA-N Synonym: 2-morpholinothiazole-4-carbaldehyde,2-morpholin-4-yl-1,3-thiazole-4-carbaldehyde,2-morpholino-1,3-thiazole-4-carbaldehyde,4-thiazolecarboxaldehyde,2-4-morpholinyl,acmc-20ahfg,4-4-formyl-1,3-thiazol-2-yl morpholine,2-morpholin-4-yl-1,3-thiazole-4-carboxaldehyde,2-morpholin-4-yl-1,3-thiazole-4-carboxaldehyd PubChem CID: 15053579 IUPAC Name: 2-morpholin-4-yl-1,3-thiazole-4-carbaldehyde SMILES: C1COCCN1C2=NC(=CS2)C=O