accessibility menu, dialog, popup

UserName

CAS RN 126533-96-8

ONNSOH

CAS RN 126533-96-8

IUPAC Name: [2-(morpholin-4-yl)-1,3-thiazol-4-yl]methanol
Synonyms: [2-(morpholin-4-yl)-1,3-thiazol-4-yl]methanol
Molecular Weight (g/mol): 200.26
Molecular Formula: C8H12N2O2S
InChi Key: QJKKFCRFBAXDQE-UHFFFAOYSA-N
SMILES: OCC1=CSC(=N1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)