accessibility menu, dialog, popup

UserName

CAS RN 1254353-37-1

IUPAC Name: 5,6-bis(octyloxy)-2,1,3-benzothiadiazole
Synonyms: 5,6-bis(octyloxy)-2,1,3-benzothiadiazole
Molecular Weight (g/mol): 392.6
Molecular Formula: C22H36N2O2S
InChi Key: GNYCNFWCQGDJRG-UHFFFAOYSA-N
SMILES: CCCCCCCCOC1=CC2=NSN=C2C=C1OCCCCCCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results