accessibility menu, dialog, popup

UserName

CAS RN 125092-42-4

H3CNHNOHNOCH3NH2

CAS RN 125092-42-4

IUPAC Name: 6-amino-1-methyl-5-(propylamino)-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 6-amino-1-methyl-5-(propylamino)-3H-pyrimidine-2,4-dione
Molecular Weight (g/mol): 198.23
Molecular Formula: C8H14N4O2
InChi Key: PXXPWRGFQVGQAZ-UHFFFAOYSA-N
SMILES: CCCNC1=C(N)N(C)C(=O)NC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results