accessibility menu, dialog, popup

UserName

NaNaH3COHOH3CHOOOPOOO

CAS RN 125-02-0

IUPAC Name: disodium 1,10-dihydroxy-9a,11a-dimethyl-1-[2-(phosphonatooxy)acetyl]-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
Synonyms: prednisolone sodium phosphate
Molecular Weight (g/mol): 484.39
Molecular Formula: C21H27Na2O8P
InChi Key: VJZLQIPZNBPASX-UHFFFAOYNA-L
SMILES: [Na+].[Na+].CC12CC(O)C3C(CCC4=CC(=O)C=CC34C)C1CCC2(O)C(=O)COP([O-])([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results