accessibility menu, dialog, popup

UserName

CAS RN 124638-53-5

IUPAC Name: tetrabromothieno[3,2-b]thiophene
Synonyms: tetrabromothieno[3,2-b]thiophene
Molecular Weight (g/mol): 455.8
Molecular Formula: C6Br4S2
InChi Key: CWCKRNKYWPNOAZ-UHFFFAOYSA-N
SMILES: BrC1=C(Br)C2=C(S1)C(Br)=C(Br)S2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results