accessibility menu, dialog, popup

UserName

NNCNNOOBrPd

CAS RN 1242081-29-3

IUPAC Name: 2,6-bis(1-benzoyl-4,5-diphenyl-4,5-dihydro-1H-imidazol-2-yl)benzen-1-ide; bromopalladiumylium
Synonyms: 2,6-bis(1-benzoyl-4,5-diphenyl-4,5-dihydroimidazol-2-yl)benzen-1-ide; bromopalladiumylium
Molecular Weight (g/mol): 912.2
Molecular Formula: C50H37BrN4O2Pd
InChi Key: KZAGTVGVDHDPDZ-UHFFFAOYNA-M
SMILES: Br[Pd+].O=C(N1C(C(N=C1C1=CC=CC(=[C-]1)C1=NC(C(N1C(=O)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results