accessibility menu, dialog, popup

UserName

CAS RN 123640-41-5

IUPAC Name: 2-bromo-6-(1H-pyrazol-1-yl)pyridine
Synonyms: 2-bromo-6-(pyrazol-1-yl)pyridine
Molecular Weight (g/mol): 224.06
Molecular Formula: C8H6BrN3
InChi Key: LPRJOZJWALGJGM-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=N1)N1C=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results