CAS RN 123387-72-4
IUPAC Name:
tert-butyl N-methyl-N-(2-oxoethyl)carbamate
Synonyms:
tert-butyl N-methyl-N-(2-oxoethyl)carbamate
Molecular Weight (g/mol):
173.21
Molecular Formula:
C8H15NO3
InChi Key:
MSWTVSDFEYSRMQ-UHFFFAOYSA-N
SMILES:
CN(CC=O)C(=O)OC(C)(C)C
1
–
1
of
1
results
1
Promotions Available
N-Boc-(methylamino)acetaldehyde, 97%
N-Boc-(methylamino)acetaldehyde, 97%
CAS: 123387-72-4 Molecular Formula: C8H15NO3 Molecular Weight (g/mol): 173.212 MDL Number: MFCD08064267 InChI Key: MSWTVSDFEYSRMQ-UHFFFAOYSA-N Synonym: n-boc-methylamino acetaldehyde,tert-butyl methyl 2-oxoethyl carbamate,tert-butyl n-methyl-n-2-oxoethyl carbamate,n-tert-butoxycarbonyl-n-methylaminoacetaldehyde,methyl-2-oxo-ethyl-carbamic acid tert-butyl ester,carbamic acid, methyl 2-oxoethyl-, 1,1-dimethylethyl ester,carbamic acid,n-methyl-n-2-oxoethyl-, 1,1-dimethylethyl ester,acmc-20a2gd,n-t-butoxycarbonylsarcosinal,n-boc-n-methyl-2-amino-acetaldehyde PubChem CID: 10725881 IUPAC Name: tert-butyl N-methyl-N-(2-oxoethyl)carbamate SMILES: CC(C)(C)OC(=O)N(C)CC=O