accessibility menu, dialog, popup

UserName

OFFFOOFFFOCu

CAS RN 123333-88-0

IUPAC Name: copper(2+) ditrifluoroacetate
Synonyms: copper(II) trifluoroacetate
Molecular Weight (g/mol): 289.58
Molecular Formula: C4CuF6O4
InChi Key: JIDMEYQIXXJQCC-UHFFFAOYSA-L
SMILES: [Cu++].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results