accessibility menu, dialog, popup

UserName

NaPtOOOOOONa

CAS RN 12325-31-4

IUPAC Name: platinum(4+) disodium hexahydroxide
Synonyms: disodium hexahydroxoplatinate
Molecular Weight (g/mol): 343.11
Molecular Formula: H6Na2O6Pt
InChi Key: LDMNYTKHBHFXNG-UHFFFAOYSA-H
SMILES: [OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Na+].[Na+].[Pt+4]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results