CAS RN 123237-62-7
IUPAC Name:
tert-butyldimethyl[(oxiran-2-yl)methoxy]silane
Synonyms:
tert-butyldimethyl(oxiran-2-ylmethoxy)silane
Molecular Weight (g/mol):
188.34
Molecular Formula:
C9H20O2Si
InChi Key:
YANSSVVGZPNSKD-UHFFFAOYNA-N
SMILES:
CC(C)(C)[Si](C)(C)OCC1CO1
1
–
1
of
1
results
1
tert-Butyldimethylsilyl (S)-Glycidyl Ether 95.0+%, TCI America™
CAS: 123237-62-7 Molecular Formula: C9H20O2Si Molecular Weight (g/mol): 188.342 MDL Number: MFCD01862148 InChI Key: YANSSVVGZPNSKD-QMMMGPOBSA-N Synonym: (S)-2-[(tert-Butyldimethylsilyloxy)methyl]oxirane, (S)-1-[(tert-Butyldimethylsilyl)oxy]-2,3-epoxypropane PubChem CID: 10954307 IUPAC Name: tert-butyl-dimethyl-[[(2S)-oxiran-2-yl]methoxy]silane SMILES: CC(C)(C)[Si](C)(C)OCC1CO1