CAS RN 123-83-1
IUPAC Name:
[2-(dimethylazaniumyl)ethyl](ethyl)azanium
Synonyms:
[2-(dimethylammonio)ethyl](ethyl)azanium
Molecular Weight (g/mol):
118.22
Molecular Formula:
C6H18N2
InChi Key:
WLNSKTSWPYTNLY-UHFFFAOYSA-P
SMILES:
CC[NH2+]CC[NH+](C)C
1
–
1
of
1
results
1
Promotions Available
N'-Ethyl-N,N-dimethylethylenediamine, 98%
N'-Ethyl-N,N-dimethylethylenediamine, 98%
CAS: 123-83-1 Molecular Formula: C6H18N2 Molecular Weight (g/mol): 118.22 MDL Number: MFCD00009034 InChI Key: WLNSKTSWPYTNLY-UHFFFAOYSA-P Synonym: n,n-dimethyl-n'-ethylethylenediamine,1,2-ethanediamine, n'-ethyl-n,n-dimethyl,2-ethylaminoethyldimethylamine,2-dimethylamino ethyl ethyl amine,ethylenediamine, n'-ethyl-n,n-dimethyl,n'-ethyl-n,n-dimethylethylenediamine,n1-ethyl-n2,n2-dimethylethane-1,2-diamine,1,2-ethanediamine, n2-ethyl-n1,n1-dimethyl,n1-ethyl-n2,n2-dimethyl-1,2-ethanediamine,n'-ethyl-n,n-dimethylethane-1,2-diamine PubChem CID: 67162 IUPAC Name: N-ethyl-N',N'-dimethylethane-1,2-diamine SMILES: CC[NH2+]CC[NH+](C)C