accessibility menu, dialog, popup

UserName

H3CNHNCH3NCH3NHCH3

CAS RN 123-67-1

IUPAC Name: 6,10-dimethyl-2,6,10,14-tetraazapentadecane
Synonyms: 6,10-dimethyl-2,6,10,14-tetraazapentadecane
Molecular Weight (g/mol): 244.43
Molecular Formula: C13H32N4
InChi Key: XRTKMBBIDHUQNP-UHFFFAOYSA-N
SMILES: CNCCCN(C)CCCN(C)CCCNC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results