accessibility menu, dialog, popup

UserName

CAS RN 12266-72-7

IUPAC Name: (1Z,5Z)-cycloocta-1,5-diene; diiodoplatinumbis(ylium)
Synonyms: 1,5-cyclooctadiene, (Z,Z)-; diiodoplatinumbis(ylium)
Molecular Weight (g/mol): 557.08
Molecular Formula: C8H12I2Pt
InChi Key: QDZCWAOYIXXIQR-PHFPKPIQSA-L
SMILES: I[Pt++]I.C1C\C=C/CC\C=C/1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results