CAS RN 122292-85-7
IUPAC Name:
7-[4-(dimethylamino)phenyl]-N-hydroxy-4,6-dimethyl-7-oxohepta-2,4-dienamide
Synonyms:
7-[4-(dimethylamino)phenyl]-N-hydroxy-4,6-dimethyl-7-oxohepta-2,4-dienamide
Molecular Weight (g/mol):
302.37
Molecular Formula:
C17H22N2O3
InChi Key:
RTKIYFITIVXBLE-UHFFFAOYNA-N
SMILES:
CC(C=C(C)C=CC(=O)NO)C(=O)C1=CC=C(C=C1)N(C)C
1
–
1
of
1
results
1
(-)-Trichostatin A 98.0+%, TCI America™
CAS: 122292-85-7 Molecular Formula: C17H22N2O3 Molecular Weight (g/mol): 302.374 InChI Key: RTKIYFITIVXBLE-LEJRBOCMSA-N Synonym: [S-(E,E)]-(-)-7-[4-(Dimethylamino)phenyl]-N-hydroxy-4,6-dimethyl-7-oxo-2,4-heptadienamide PubChem CID: 13507024 IUPAC Name: (2E,4E,6S)-7-[4-(dimethylamino)phenyl]-N-hydroxy-4,6-dimethyl-7-oxohepta-2,4-dienamide SMILES: CC(C=C(C)C=CC(=O)NO)C(=O)C1=CC=C(C=C1)N(C)C