accessibility menu, dialog, popup

UserName

H3CH3CCH3OONHBr

CAS RN 1222709-30-9

IUPAC Name: tert-butyl N-[4-(bromomethyl)cyclohexyl]carbamate
Synonyms: tert-butyl N-[4-(bromomethyl)cyclohexyl]carbamate
Molecular Weight (g/mol): 292.22
Molecular Formula: C12H22BrNO2
InChi Key: JPJCPRFAUVDSEA-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1CCC(CBr)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results