accessibility menu, dialog, popup

UserName

CAS RN 1218791-15-1

BOOH3CCH3H3CH3CNF

CAS RN 1218791-15-1

IUPAC Name: 2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Synonyms: 2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Molecular Weight (g/mol): 247.08
Molecular Formula: C13H15BFNO2
InChi Key: BDIHSSFYJYXWRV-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C(F)C(=CC=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results