CAS RN 1217891-71-8
IUPAC Name:
9-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
Synonyms:
9-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
Molecular Weight (g/mol):
307.2
Molecular Formula:
C19H22BNO2
InChi Key:
AVCKYKVGQYBADD-UHFFFAOYSA-N
SMILES:
CN1C2=C(C=CC=C2)C2=C1C=CC(=C2)B1OC(C)(C)C(C)(C)O1
1
–
1
of
1
results
1
Promotions Available
N-Methylcarbazole-3-boronic acid pinacol ester, 97%
N-Methylcarbazole-3-boronic acid pinacol ester, 97%
CAS: 1217891-71-8 Molecular Formula: C19H22BNO2 Molecular Weight (g/mol): 307.20 MDL Number: MFCD16294552 InChI Key: AVCKYKVGQYBADD-UHFFFAOYSA-N Synonym: 9-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-9h-carbazole,9-methyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-9h-carbazole,9-methylcarbazole-3-boronic acid pinacol ester,9-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl carbazole,amtb254,9-methyl-9h-carbazole-3-boronic acid pinacol ester,3-4,4,5,5-tetramethyl-1,3,2-dioxaborole-2-yl-9-methyl-9h-carbazole PubChem CID: 56924967 SMILES: CN1C2=C(C=CC=C2)C2=C1C=CC(=C2)B1OC(C)(C)C(C)(C)O1