accessibility menu, dialog, popup

UserName

NH3H2O

CAS RN 12168-30-8

IUPAC Name: amine hydrate
Synonyms: ammonium hydroxide
Molecular Weight (g/mol): 35.05
Molecular Formula: H5NO
InChi Key: VHUUQVKOLVNVRT-UHFFFAOYSA-N
SMILES: N.O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results