CAS RN 1211578-46-9
IUPAC Name:
6-chloro-2-fluoropyridine-3-carboxylic acid
Synonyms:
6-chloro-2-fluoropyridine-3-carboxylic acid
Molecular Weight (g/mol):
175.54
Molecular Formula:
C6H3ClFNO2
InChi Key:
GSSVAHBRTISODE-UHFFFAOYSA-N
SMILES:
OC(=O)C1=C(F)N=C(Cl)C=C1
1
–
1
of
1
results
1
Promotions Available
6-Chloro-2-fluoronicotinic acid, 95%
6-Chloro-2-fluoronicotinic acid, 95%
CAS: 1211578-46-9 Molecular Formula: C6H3ClFNO2 Molecular Weight (g/mol): 175.54 MDL Number: MFCD16610397 InChI Key: GSSVAHBRTISODE-UHFFFAOYSA-N Synonym: 6-chloro-2-fluoronicotinic acid,6-chloro-2-fluoro nicotinic acid,6-chloro-2-fluoronicotinicacid,6-chloro-2-fluoropyridin-3-carboxylic acid,6-chloro-2-fluoro-pyridine-3-carboxylic acid PubChem CID: 71464004 IUPAC Name: 6-chloro-2-fluoropyridine-3-carboxylic acid SMILES: OC(=O)C1=C(F)N=C(Cl)C=C1