accessibility menu, dialog, popup

UserName

CAS RN 121062-08-6

H3C(S)HNCH3OONH(S)ONH(S)NHO(S)NHNHO(S)NNH2NH2NHO(R)HNO(S)NHNNHONH2O

CAS RN 121062-08-6

IUPAC Name: (3S,6S,9R,12S,15S,23S)-9-benzyl-6-{3-[(diaminomethylidene)amino]propyl}-15-[(2S)-2-acetamidohexanamido]-12-[(1H-imidazol-5-yl)methyl]-3-[(1H-indol-3-yl)methyl]-2,5,8,11,14,17-hexaoxo-1,4,7,10,13,18-hexaazacyclotricosane-23-carboxamide
Synonyms: melanotan II
Molecular Weight (g/mol): 1024.2
Molecular Formula: C50H69N15O9
InChi Key: JDKLPDJLXHXHNV-MFVUMRCOSA-N
SMILES: CCCC[C@H](NC(C)=O)C(=O)N[C@H]1CC(=O)NCCCC[C@H](NC(=O)[C@H](CC2=CNC3=CC=CC=C23)NC(=O)[C@H](CCCN=C(N)N)NC(=O)[C@@H](CC2=CC=CC=C2)NC(=O)[C@H](CC2=CN=CN2)NC1=O)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)