accessibility menu, dialog, popup

UserName

CAS RN 12075-68-2

IUPAC Name: chlorodiethylalumane; dichloro(ethyl)alumane
Synonyms: ethylaluminium sesquichloride
Molecular Weight (g/mol): 247.5
Molecular Formula: C6H15Al2Cl3
InChi Key: LDXMUMPEDOQULK-UHFFFAOYSA-K
SMILES: CC[Al](Cl)Cl.CC[Al](Cl)CC
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)