accessibility menu, dialog, popup

UserName

CAS RN 12069-89-5

IUPAC Name: (molybdeniomethylidene)molybdenum
Synonyms: (molybdeniomethylidene)molybdenum
Molecular Weight (g/mol): 203.91
Molecular Formula: CMo2
InChi Key: QIJNJJZPYXGIQM-UHFFFAOYSA-N
SMILES: [Mo]=C=[Mo]
Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results