accessibility menu, dialog, popup

UserName

BrSI

CAS RN 1206544-88-8

IUPAC Name: 4-bromo-12-iodo-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Synonyms: 4-bromo-12-iodo-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Molecular Weight (g/mol): 389.05
Molecular Formula: C12H6BrIS
InChi Key: HMECZSLDNCAHAV-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(SC3=C2C=C(I)C=C3)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results