accessibility menu, dialog, popup

UserName

CAS RN 12035-51-7

NiSS

CAS RN 12035-51-7

IUPAC Name: disulfanylidenenickel
Synonyms: disulfanylidenenickel
Molecular Weight (g/mol): 122.81
Molecular Formula: NiS2
InChi Key: HNDMNBUITRNJKK-UHFFFAOYSA-N
SMILES: S=[Ni]=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results