accessibility menu, dialog, popup

UserName

NTa

CAS RN 12033-62-4

IUPAC Name: tantalioamine
Synonyms: tantalum nitride
Molecular Weight (g/mol): 194.96
Molecular Formula: NTa
InChi Key: MZLGASXMSKOWSE-UHFFFAOYSA-N
SMILES: N#[Ta]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results